Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibutylboron trifluoromethanesulfonate 1 M in dichloromethane

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

37899-0250 378991000
TS37899-0250EA 93 USD InStock
TS37899-0250 TS378991000
Dibutylboron trifluoromethanesulfonate 1 M in dichloromethane
Dibutylboron trifluoromethanesulfonate


Formula: C₉H₁₈BF₃O₃S
Density: 1.271 g/cm³
Flash Pt: 27 °C
Storage Temperature: Freezer
MDL Number: MFCD00009669
CAS Number: 60669-69-4
UN: 2920
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to orange Solution
Strength 0.9 to 1.1 M

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty