Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium sulfite 90%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

44021-0025 AC44021-0010 AC44021-2500
TS44021-0025EA 246 USD InStock
TS44021-0025 TS44021-0010 TS44021-2500
Potassium sulfite 90%, pure
Potassium sulfite


Formula: K₂SO₃
Storage Temperature: Ambient
MDL Number: MFCD00011387
CAS Number: 10117-38-1
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration Iodimetric ≥88 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty