Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Naproxen sodium 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

43671-0250 43671-0050
TS43671-0250EA 269 USD InStock
TS43671-0250 TS43671-0050
Naproxen sodium 98%
Naproxen sodium


Formula: C₁₄H₁₃NaO₃
Melting Pt: 259…262 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00058507
CAS Number: 26159-34-2
Order Now
Specification Test Results
Appearance Colorless to light yellow Powder
Infrared spectrum Conforms
Specific optical rotation -14.7° to -17.0° (20°C, 589 nm) (c=5, 1N NaOH)
Titration with HClO4 98.0 to 102.0 %
Free acid ≤1.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty