Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium hydrogen-L-tartrate 98+% for analysis

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

38942-2500 AC38942-0010
TS38942-2500EA 58 USD InStock
TS38942-2500 TS38942-0010
Sodium hydrogen-L-tartrate 98+% for analysis
Sodium hydrogen-L-tartrate


Formula: C₄H₅NaO₆
MW: 172.07 g/mol
Melting Pt: 253 °C
Storage Temperature: Ambient
MDL Number: MFCD00065393
CAS Number: 526-94-3
Order Now
Specification Test Results
Appearance White Powder
pH 3.0 to 3.6 (5 % solution)
Titration with NaOH ≥98.0 %
Chloride (Cl) ≤0.02 %
Sulfate (SO4) ≤0.02 %
Heavy metals (as Pb) ≤0.002 %
Insoluble matter ≤0.03 % (in water)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty