Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Propynylmagnesium bromide 0.5 M in THF, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

42607-1000 AC42607-8000
TS42607-1000EA 136 USD InStock
TS42607-1000 TS42607-8000
1-Propynylmagnesium bromide 0.5 M in THF, AcroSeal™
1-Propynylmagnesium bromide


Formula: C₃H₃BrMg
Boiling Pt: 65…67 °C (760 mmHg)
Density: 0.94 g/cm³
Flash Pt: –1 °C
Storage Temperature: Ambient
MDL Number: MFCD00216588
CAS Number: 16466-97-0
UN: 3399
ADR: 4.3,I
Order Now
Specification Test Results
Appearance Clear colorless to yellow Solution (May develop some turbidity or precipitate)
Titration Grignard 0.45 to 0.65 M
Refractive index 1.5270 to 1.5310 (20°C, 589 nm)
GC ≥95 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty