Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butylphosphonic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

32152-0050 32152-0010
TS32152-0050EA 188 USD InStock
TS32152-0050 TS32152-0010
tert-Butylphosphonic acid 98%
tert-Butylphosphonic acid


Formula: C₄H₁₁O₃P
Boiling Pt: 67 °C (760 mmHg)
Melting Pt: –113 °C
Density: 0.677 g/cm³
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00012296
CAS Number: 4923-84-6
EINECS: 236-261-4
UN: 3295
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3935 to 1.3955 (20°C, 589 nm)
Assay ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty