Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Oleanolic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45612-0250 45612-0050
TS45612-0250EA 252 USD InStock
TS45612-0250 TS45612-0050
Oleanolic acid 97%
Oleanolic acid


Formula: C₃₀H₄₈O₃
MW: 456.71 g/mol
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064914
CAS Number: 508-02-1
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Water ≤5 %
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty