Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(-)-Glutathione (oxidised form) hydrate 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

34384-0050 34384-0250 34384-0010
TS34384-0050EA 225 USD InStock
TS34384-0050 TS34384-0250 TS34384-0010
L(-)-Glutathione (oxidised form) hydrate 95%
L(-)-Glutathione (oxidised form) hydrate


Formula: C₂₀H₃₂N₆O₁₂S₂·xH₂O
Melting Pt: 178 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00150701
CAS Number: 27025-41-8
EINECS: 248-170-7
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤8 % (K.F.)
Sulfated ash ≤0.1 % (800°C)
HPLC ≥94 %
Specific optical rotation -92° to -106° (20°C, 589 nm) (c=2, H2O)
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty