Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phosphomolybdic acid 20% (w/w) in ethanol

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

36862-1000 36862-5000
TS36862-1000EA 78 USD InStock
TS36862-1000 TS36862-5000
Phosphomolybdic acid 20% (w/w) in ethanol
Phosphomolybdic acid solutions


Formula: H₃[P(Mo₃O₁₀)₄]
Melting Pt: 78…90 °C
Density: 0.94 g/cm³
Flash Pt: 5 °C
Storage Temperature: Ambient
MDL Number: MFCD00011341
CAS Number: 51429-74-4
UN: 2924
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear yellow to green Solution
Titration with NaOH 19 to 21 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty