Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium oxide acidic Brockmann Grade I 40-300 µm, 60A for chromatography

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

36575-0010 AC36575-0025 AC36575-0100
TS36575-0010EA 108 USD InStock
TS36575-0010 TS36575-0025 TS36575-0100
Aluminium oxide acidic Brockmann Grade I 40-300 µm, 60A for chromatography
Aluminium oxide acidic


Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C (760 mmHg)
Melting Pt: 2030 °C
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
Order Now
Specification Test Results
Appearance White to off-white Powder
Particle size ≤5 % (<40 µm)
Specific surface area 185 to 225 m²/g
pH 4.0 to 5.0 (10% aqueous suspension)
Sodium oxide (Na2O) ≤0.4 %
Silicon dioxide (SiO2) ≤0.03 %
Iron(III)oxide (Fe2O3) ≤0.03 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty