Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethanol ≥95% ACS for spectrophotometry (190 Proof)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

61511-0010 61511-0040
TS61511-0010EA 185 USD InStock
TS61511-0010 TS61511-0040
Ethanol ≥95% ACS for spectrophotometry (190 Proof)
Ethanol


Formula: C₂H₅OH
Boiling Pt: 78 °C
Flash Pt: 17 °C
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3605 to 1.3625 (25 °C, 589 nm)
Assay ≥95.0 % v/v
Solubility (in water) Passes test
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 Passes test
Titrable acid ≤0.0005 meq/g
Titrable base ≤0.0002 meq/g
KMnO4-reducing substance Passes test

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty