Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Silicone oil, 475 -525 mm²/s (25 °C) cP for melting point and boiling point apparatuses

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

16385-0010 16385-0025 16385-5000
TS16385-0010EA 130 USD InStock
TS16385-0010 TS16385-0025 TS16385-5000
Silicone oil, 475 -525 mm²/s (25 °C) cP for melting point and boiling point apparatuses
Silicone oil


Melting Pt: –50 °C
Density: 0.96
Flash Pt: 300 °C
MDL Number: MFCD00132673
CAS Number: 68083-14-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty