Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate ≥98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43907-0010 43907-0050
TS43907-0010EA 59 USD InStock
TS43907-0010 TS43907-0050
Chloro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate ≥98%
Chloro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate


Formula: C₅H₁₂ClF₆N₂P
Storage Temperature: Freezer
MDL Number: MFCD01862891
CAS Number: 94790-35-9
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 100°C to 106°C
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty