Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Clobetasol propionate ≥97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

46682-0010 46682-0050
TS46682-0010EA 58 USD InStock
TS46682-0010 TS46682-0050
Clobetasol propionate ≥97%
Clobetasol propionate


Formula: C₂₅H₃₂ClFO₅
MDL Number: MFCD00058499
CAS Number: 25122-46-7
EINECS: 246-634-3
Order Now
Specification Test Results
Appearance Crystalline powder, White to almost white
Infrared spectrum Conforms
Melting point 196.0°C to 200.0°C
HPLC >=97.0 %
Loss on drying =<0.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty