Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Allyltriphenylphosphonium bromide 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

10304-0250 10304-1000
TS10304-0250EA 23 USD InStock
TS10304-0250 TS10304-1000
Allyltriphenylphosphonium bromide 99%
Allyltriphenylphosphonium bromide


Formula: C₂₁H₂₀BrP
Melting Pt: 222…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00011808
CAS Number: 1560-54-9
EINECS: 216-332-6
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Water ≤12 % (K.F.)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty