Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-(Diethylamino)-4-methylcoumarin 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

11405-0050 11405-1000 11405-5000
TS11405-0050EA 39 USD InStock
TS11405-0050 TS11405-1000 TS11405-5000
7-(Diethylamino)-4-methylcoumarin 99%
7-(Diethylamino)-4-methylcoumarin


Formula: C₁₄H₁₇NO₂
Melting Pt: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00006864
CAS Number: 91-44-1
EINECS: 202-068-9
Order Now
Specification Test Results
Appearance Light beige to light purple Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 73 °C
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty