Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl chloroformate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

12653-0025 12653-0050 12653-1000 12653-5000
TS12653-0025EA 358 USD InStock
TS12653-0025 TS12653-0050 TS12653-1000 TS12653-5000
Methyl chloroformate 99%
Methyl chloroformate


Formula: C₂H₃ClO₂
Boiling Pt: 70…72 °C (760 mmHg)
Melting Pt: –61 °C
Density: 1.223 g/cm³
Flash Pt: 5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000639
CAS Number: 79-22-1
EINECS: 201-187-3
UN: 1238
ADR: 6.1,
Merck Index: 1206123
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3860 to 1.3880 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty