Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methylene blue trihydrate 95%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12675-0010 12675-1000 12675-2500
TS12675-0010EA 663 USD InStock
TS12675-0010 TS12675-1000 TS12675-2500
Methylene blue trihydrate 95%, pure
Methylene blue trihydrate


Formula: C₁₆H₁₈ClN₃S·3H₂O
MW: 373.9 g/mol
Melting Pt: 100…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS Number: 7220-79-3
EINECS: 200-515-2
Order Now
Specification Test Results
Appearance Dark green Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥94 % (on dry substance)
Loss on drying ≤15 % (105°C)
Color scale ≤20 APHA
Acidity ≤0.001 meq/g
Alkalinity ≤0.002 meq/g
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Tin (Sn) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Heavy metals (as Pb) ≤1 ppm
Halogenated compounds ≤0.001 % (as Cl)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty