Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Propanedithiol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

13141-0500 13141-2500
TS13141-0500EA 120 USD InStock
TS13141-0500 TS13141-2500
1,3-Propanedithiol 98%
1,3-Propanedithiol


Formula: C₃H₈S₂
Boiling Pt: 169 °C (760 mmHg)
Melting Pt: –79 °C
Density: 1.07 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00004904
CAS Number: 109-80-8
EINECS: 203-706-9
UN: 1228
ADR: 3,III
Merck Index: 1207984
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5380 to 1.5420 (20°C, 589 nm)
GC ≥97.5 %
Density (20°C) 1.038 to 1.048 g/ml

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty