Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Sulphosalicylic acid dihydrate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

13286-0010 13286-2500
TS13286-0010EA 153 USD InStock
TS13286-0010 TS13286-0025 TS13286-2500
5-Sulphosalicylic acid dihydrate 98%
5-Sulphosalicylic acid dihydrate


Formula: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
MW: 254.22 g/mol
Melting Pt: 224
Flash Pt: 150
Storage Temperature: Ambient
MDL Number: MFCD00149540
CAS Number: 5965-83-3
EINECS: 202-555-6
UN: 2585
ADR: 8 III
Merck Index: 13,09054
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69e00688850e885438e2fdd9a62f6d564ebf6ae8&vid=69e00688850e885438e2fdd9a62f6d564ebf6ae8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 106 to 110 °C
Water 14 to 16 % (K.F.) Typical
Titration with NaOH ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty