Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Safranine O 95%, pure, high purity biological stain

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

14664-0250 14664-1000
TS14664-0250EA 69 USD InStock
TS14664-0250 TS14664-1000
Safranine O 95%, pure, high purity biological stain
Safranine O


Formula: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6
EINECS: 207-518-8
Order Now
Specification Test Results
Appearance Green to red-brown Powder
Infrared spectrum Conforms
Loss on drying ≤10 %
Dye content ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty