Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium hydroxymethanesulphinate dihydrate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

14919-0010 14919-0050 14919-0250
TS14919-0010EA 69 USD InStock
TS14919-0010 TS14919-0050 TS14919-0250
Sodium hydroxymethanesulphinate dihydrate 98%
Sodium hydroxymethanesulphinate dihydrate


Formula: CH₃NaO₃S·2H₂O
MW: 154.12 g/mol
Melting Pt: 63…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00150598
CAS Number: 6035-47-8
EINECS: 205-739-4
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Iodometric ≥97.5 %
pH 9.5 to 10.5 (10 % solution)
Alkalinity 1 to 2.5 % (as Na2CO3)
Iron (Fe) ≤10 ppm
Zinc (Zn) ≤30 ppm

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty