Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromocresol purple, pure indicator grade

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

15133-0010 15133-0050 15133-0250 15133-1400
TS15133-0010EA 1193 USD InStock
TS15133-0010 TS15133-0050 TS15133-0250 TS15133-1400
Bromocresol purple, pure indicator grade
Bromocresol purple


Formula: C₂₁H₁₆Br₂O₅S
MW: 540.23 g/mol
Melting Pt: 242 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011681
CAS Number: 115-40-2
EINECS: 204-087-8
Merck Index: 13,01371
Order Now
Specification Test Results
Appearance Light pink to pink-orange Powder
Infrared spectrum Conforms
Solubility (0.1 % in 50 % ethanol) Clear
UV (20 mg/l in methanol) Complies
Visual transition interval from pH 5.2 (yellow) to pH 6.8 (purple)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty