Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

TBPE (=tetrabromophenolphthalein ethyl ester potassium salt), pure indicator grade

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

15158-0010 15158-0050 15158-0250
TS15158-0010EA 111 USD InStock
TS15158-0010 TS15158-0050 TS15158-0250
TBPE (=tetrabromophenolphthalein ethyl ester potassium salt), pure indicator grade
TBPE (=tetrabromophenolphthalein ethyl ester potassium salt)


Formula: C₂₂H₁₄Br₄O₄
MW: 700.06 g/mol
Melting Pt: 270…276 °C
Storage Temperature: Ambient
MDL Number: MFCD00011662
CAS Number: 62637-91-6
Order Now
Specification Test Results
Appearance Blue to blue-black Crystalline powder or granules
Infrared spectrum Conforms
Specific extinction E (1%/1cm) ≥660 (600 nm, ethanol)
Specific optical rotation -32° to -38° (20°C, 589 nm) (c=1 in EtOH)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty