Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,5-Diazobicyclo[4.3.0]-5-nonene 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15961-0250 15961-1000
TS15961-0250EA 82 USD InStock
TS15961-0250 TS15961-1000
1,5-Diazobicyclo[4.3.0]-5-nonene 98%
1,5-Diazobicyclo[4.3.0]-5-nonene


Formula: C₇H₁₂N₂
MW: 124.19 g/mol
Boiling Pt: 110
Density: 1 g/cm³
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00005554
CAS Number: 3001-72-7
EINECS: 221-087-3
UN: 3267
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5190 to 1.5210 (20°C, 589 nm)
Water ≤1 %
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty