Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Dimethylamino)benzaldehyde 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

16876-0025 16876-0250 16876-1000 16876-5000
TS16876-0025EA 680 USD InStock
TS16876-0025 TS16876-0250 TS16876-1000 TS16876-5000
4-(Dimethylamino)benzaldehyde 99+%
4-(Dimethylamino)benzaldehyde


Formula: (CH₃)₂NC₆H₄CHO
MW: 149.19 g/mol
Boiling Pt: 176-177
Melting Pt: 72
Flash Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS Number: 100-10-7
EINECS: 202-819-0
Merck Index: 13,03257
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=a978f8cb2be9fb6dfa0aa34ce0038883c864820b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to light green Crystalline powder
Infrared spectrum Conforms
Melting point 72 to 75 °C
GC ≥99.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty