Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrophenyl chloroformate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

17080-0050 17080-0250 17080-1000 17080-5000
TS17080-0050EA 51 USD InStock
TS17080-0050 TS17080-0250 TS17080-1000 TS17080-5000
4-Nitrophenyl chloroformate 97%
4-Nitrophenyl chloroformate


Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Boiling Pt: 159
Melting Pt: 73…81 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007321
CAS Number: 7693-46-1
EINECS: 231-706-9
UN: 2923
ADR: 8 II
Order Now
Specification Test Results
Appearance White to yellow to light beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 77 to 82 °C
Titration Argentometric ≥96.0 % (ex Cl)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty