Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(+)-Dimethyl-L-tartrate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

17249-0250 17249-1000 17249-5000
TS17249-0250EA 32 USD InStock
TS17249-0250 TS17249-1000 TS17249-5000
(+)-Dimethyl-L-tartrate 99%
(+)-Dimethyl-L-tartrate


Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Boiling Pt: 245
Melting Pt: 48…50 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00064437
CAS Number: 608-68-4
EINECS: 210-166-8
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
GC ≥98.5 %
Specific optical rotation +20° to +22° (20°C, 589 nm) (10% in water)
Elemental Analysis 5.22 to 5.65 % (N)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty