Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bicine 99+% for analysis

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

17265-0010 17265-1000 17265-2500
TS17265-0010EA 525 USD InStock
TS17265-0010 TS17265-1000 TS17265-2500
Bicine 99+% for analysis
Bicine


Formula: C₆H₁₃NO₄
MW: 163.17 g/mol
Melting Pt: 190…192 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004295
CAS Number: 150-25-4
EINECS: 205-755-1
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals or needles
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
Sulfated ash ≤0.1 %
UV at 250 nm A: ≤0.05
pH 4.0 to 5.5 (10 % at 20°C)
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty