Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethyl orthobenzoate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

17292-0100 17292-0500
TS17292-0100EA 39 USD InStock
TS17292-0100 TS17292-0500
Trimethyl orthobenzoate 97%
Trimethyl orthobenzoate


Formula: C₁₀H₁₄O₃
MW: 182.22
Boiling Pt: 87…88 °C (7 mmHg)
Flash Pt: 82 °C
MDL Number: MFCD00008474
CAS Number: 707-07-3
EINECS: 211-897-5
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4910 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty