Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

o-Cresolphthalein complexone, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

18244-0050 18244-0250 18244-1000
TS18244-0050EA 74 USD InStock
TS18244-0050 TS18244-0250 TS18244-1000
o-Cresolphthalein complexone, pure
o-Cresolphthalein complexone


Formula: C₃₂H₃₂N₂O₁₂
MW: 636.62 g/mol
Melting Pt: 181 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005911
CAS Number: 2411-89-4
EINECS: 219-318-8
Order Now
Specification Test Results
Appearance Off-white to pink-beige Powder
Infrared spectrum Conforms
UV spectrum Complies
Loss on drying ≤10 % (1 g, 105°C)
Sulfated ash ≤0.5 %
UV Lambda max 573 to 579 nm (in 0.1N NaOH)
Suitability (as complexom. indicator) Passes test

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty