Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fluorescein diacetate 97%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

19166-0050 19166-0250 19166-1000
TS19166-0050EA 83 USD InStock
TS19166-0050 TS19166-0250 TS19166-1000
Fluorescein diacetate 97%, pure
Fluorescein diacetate


Formula: C₂₄H₁₆O₇
Melting Pt: 200…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00005062
CAS Number: 596-09-8
EINECS: 209-877-6
Order Now
Specification Test Results
Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 200 to 205 °C
Sulfated ash ≤0.1 %
HPLC ≥96.0 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty