Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

19845-0250 19845-1000 19845-5000
TS19845-0250EA 51 USD InStock
TS19845-0250 TS19845-1000 TS19845-5000
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid) 98%
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid)


Formula: C₆H₈O₄
Melting Pt: 92…96 °C
Flash Pt: >365 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006638
CAS Number: 2033-24-1
EINECS: 217-992-8
UN: 3077
Order Now
Specification Test Results
Appearance White to beige to light brown Powder or crystals
Infrared spectrum Conforms
Melting point 92 to 96 °C
Water ≤0.1 % (K.F.)
Titration with NaOH ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty