Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyronin G, high purity biological stain

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

20010-0010 20010-0050 20010-0250
TS20010-0010EA 47 USD InStock
TS20010-0010 TS20010-0050 TS20010-0250
Pyronin G, high purity biological stain
Pyronin G


Formula: C₁₇H₁₉ClN₂O
MW: 302.8 g/mol
Melting Pt: 250…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00011725
CAS Number: 92-32-0
EINECS: 202-147-8
Merck Index: 1208190
Order Now
Specification Test Results
Appearance Dark green Crystalline powder
Infrared spectrum Conforms
Dye content ≥50 %
UV Lambda max 546 to 551 nm (in 50% ethanol)
Absorption Ratio P-15/P+15 1.23 to 1.7

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty