Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dimethylmethyleneammonium iodide 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

20101-0100 20101-0500
TS20101-0100EA 111 USD InStock
TS20101-0100 TS20101-0500
N,N-Dimethylmethyleneammonium iodide 97%
N,N-Dimethylmethyleneammonium iodide


Formula: C₃H₈IN
MW: 185.01 g/mol
Melting Pt: 219 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011810
CAS Number: 33797-51-2
Order Now
Specification Test Results
Appearance Cream to yellow Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty