Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trioctylphosphine oxide 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

20760-0250 20760-1000 20760-5000
TS20760-0250EA 62 USD InStock
TS20760-0250 TS20760-1000 TS20760-5000
Trioctylphosphine oxide 99%
Trioctylphosphine oxide


Formula: C₂₄H₅₁OP
Boiling Pt: 201…202 °C (2 mmHg)
Melting Pt: 50…55 °C
Flash Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00002083
CAS Number: 78-50-2
EINECS: 201-121-3
Order Now
Specification Test Results
Appearance White to light yellow Powder or crystals or flakes and/or chunks
Infrared spectrum Conforms
Melting point 50 to 55 °C
GC ≥98.5 % (including isomer)
Isomer ≤6 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty