Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chlorophenol red sodium salt, water soluble, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

21631-0100 21631-0500
TS21631-0100EA 90 USD InStock
TS21631-0100 TS21631-0500
Chlorophenol red sodium salt, water soluble, pure
Chlorophenol red sodium salt


Formula: C₁₉H₁₂Cl₂O₅S
MW: 445.24 g/mol
Melting Pt: 267 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00151199
CAS Number: 123333-64-2
Order Now
Specification Test Results
Appearance Dark brown Crystalline powder
Infrared spectrum Conforms
Solubility (0.1 % in water) Clear
Specific extinction E (1%/1cm) (575 nm, 0.1 N NaOH) ca. 1200
Visual transition interval from pH 4.6 (yellow) to pH 7.0 (purple)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty