Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-Dicyclohexano-18-crown-6 (mixture of syn-cis and anti-cis isomers) 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

27752-0025 27752-0100
TS27752-0025EA 58 USD InStock
TS27752-0025 TS27752-0100
cis-Dicyclohexano-18-crown-6 (mixture of syn-cis and anti-cis isomers) 98%
cis-Dicyclohexano-18-crown-6 (mixture of syn-cis and anti-cis isomers)


Formula: C₂₀H₃₆O₆
Boiling Pt: 181…183 °C
Melting Pt: 47…53 °C
Storage Temperature: Refrigerator
CAS Number: 16069-36-6
EINECS: 240-216-4
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to light beige Crystalline solid
Infrared spectrum Conforms
Melting point 47 to 53 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty