Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-tert-Butylcalix[4]arene 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

29022-0050 29022-0250
TS29022-0050EA 100 USD InStock
TS29022-0050 TS29022-0250
4-tert-Butylcalix[4]arene 99%
4-tert-Butylcalix[4]arene


Formula: C₄₄H₅₆O₄
MW: 648.93 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00066280
CAS Number: 60705-62-6
Order Now
Specification Test Results
Appearance White to light beige Crystalline powder or crystals or flakes
Infrared spectrum Conforms
HPLC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty