Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichloro(methyl)phosphine 90% tech.

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

31024-0100 31024-0500
TS31024-0100EA 207 USD InStock
TS31024-0100 TS31024-0500
Dichloro(methyl)phosphine 90% tech.
Dichloro(methyl)phosphine


Formula: CH₃Cl₂P
Boiling Pt: 81…82 °C (760 mmHg)
Density: 1.3 g/cm³
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00013616
CAS Number: 676-83-5
EINECS: 211-631-8
UN: 2845
ADR: 4.2,I
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4945 to 1.4985 (20°C, 589 nm)
GC ≥88 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty