Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxyphenyl chloroformate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

31457-0050 31457-0250
TS31457-0050EA 70 USD InStock
TS31457-0050 TS31457-0250
4-Methoxyphenyl chloroformate 98%
4-Methoxyphenyl chloroformate


Formula: C₇H₄BrClO₂
Boiling Pt: 49 °C (.2 mmHg)
Density: 1.25 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013258
CAS Number: 7693-41-6
UN: 3277
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5220 to 1.5260 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty