Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(2-diphenylphosphinoethyl)phenylphosphine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

31682-0010 31682-0050
TS31682-0010EA 65 USD InStock
TS31682-0010 TS31682-0050
Bis(2-diphenylphosphinoethyl)phenylphosphine 97%
Bis(2-diphenylphosphinoethyl)phenylphosphine


Formula: C₃₄H₃₃P₃
Melting Pt: 126…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00003048
CAS Number: 23582-02-7
Order Now
Specification Test Results
Appearance White to off-white Powder and/or chunks
Infrared spectrum Conforms
Melting point 126 to 132 °C
Elemental Analysis 74.1 to 78.7 % (C)

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty