Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Xylenol orange tetrasodium salt, water soluble, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

21122-0010 21122-0050 21122-0100 21122-0500
TS21122-0010EA 52 USD InStock
TS21122-0010 TS21122-0050 TS21122-0100 TS21122-0500
Xylenol orange tetrasodium salt, water soluble, pure
Xylenol orange tetrasodium salt


Formula: C₃₁H₂₈N₂Na₄O₁₃S
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00044293
CAS Number: 3618-43-7
EINECS: 222-805-8
Order Now
Specification Test Results
Appearance Red-brown to dark brown Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
Loss on drying ≤10.0 % (110°C, 1 h)
UV Lambda max 575 to 585 nm (0.1N NaOH)
Suitability (for Zn titration) Passes test

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty