Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Diethylamino)sulphur trifluoride 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

21611-0050 21611-0250 21611-1000
TS21611-0050EA 120 USD InStock
TS21611-0050 TS21611-0250 TS21611-1000
(Diethylamino)sulphur trifluoride 95%
(Diethylamino)sulphur trifluoride


Formula: C₄H₁₀F₃NS
MW: 161.19 g/mol
Boiling Pt: 30…32 °C (3 mmHg)
Density: 1.316 g/cm³
Flash Pt: 23 °C
Storage Temperature: Freezer
MDL Number: MFCD00000363
CAS Number: 38078-09-0
EINECS: 253-771-2
UN: 2920
ADR: 8 I
Order Now
Specification Test Results
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4160 (20°C, 589 nm)
NMR (1H-NMR) ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty