Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,3,3-Tetramethylguanidine 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

22129-0020 22129-1000 22129-5000
TS22129-0020EA 560 USD InStock
TS22129-0020 TS22129-1000 TS22129-5000
1,1,3,3-Tetramethylguanidine 99%
1,1,3,3-Tetramethylguanidine


Formula: C₅H₁₃N₃
MW: 115.18 g/mol
Boiling Pt: 160…162 °C (760 mmHg)
Melting Pt: –30 °C
Density: 0.91 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00008323
CAS Number: 80-70-6
EINECS: 201-302-7
UN: 2920
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4665 to 1.4695 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty