Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

29149-0050 29149-0250 29149-1000
TS29149-0050EA 45 USD InStock
TS29149-0050 TS29149-0250 TS29149-1000
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) 96%
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)


Formula: C₇H₁₄B₂ClF₉N₂
MW: 354.26 g/mol
Melting Pt: 260 °C (dec.)
Flash Pt: >54 °C
Storage Temperature: Ambient
MDL Number: MFCD00142607
CAS Number: 140681-55-6
UN: 3088
ADR: 4.2 II
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Titration Iodometric ≥95 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty