Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl isocyanoacetate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

29955-0050 29955-0250 29955-1000
TS29955-0050EA 104 USD InStock
TS29955-0050 TS29955-0250 TS29955-1000
Ethyl isocyanoacetate 97%
Ethyl isocyanoacetate


Formula: C₅H₇NO₂
MW: 113.12 g/mol
Boiling Pt: 194…196 °C (760 mmHg)
Density: 1.03 g/cm³
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000007
CAS Number: 2999-46-4
EINECS: 221-077-9
UN: 2810
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear amber to dark brown Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4250 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty