Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

EDTA tripotassium salt dihydrate 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

30160-0025 30160-1000 30160-5000
TS30160-0025EA 2134 USD InStock
TS30160-0025 TS30160-1000 TS30160-5000
EDTA tripotassium salt dihydrate 99+%
EDTA tripotassium salt dihydrate


Formula: C₁₀H₁₃K₃N₂O₈·2H₂O
MW: 442.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149672
CAS Number: 65501-24-8
EINECS: 241-543-5
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Complexometric ≥99.0 % (on dry substance)
Water ≤9 % (K.F.)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty