Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Mercaptobenzothiazole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

33205-0010 33205-0050 33205-0250 33205-1000
TS33205-0010EA 62 USD InStock
TS33205-0010 TS33205-0050 TS33205-0250 TS33205-1000
2-Mercaptobenzothiazole 97%
2-Mercaptobenzothiazole


Formula: C₇H₅NS₂
Melting Pt: 172…182 °C
Flash Pt: 243 °C
Storage Temperature: Ambient
MDL Number: MFCD00005781
CAS Number: 149-30-4
EINECS: 205-736-8
UN: 3077
ADR: 9,III
Merck Index: 13,05893
Order Now
Specification Test Results
Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 177 to 182 °C
Solubility (2.5% in DMSO) Clear colourless to yellow solution
Titration with NaOH 96.0 to 104.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty