Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Di-tert-butyl iminodicarboxylate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36966-0050 36966-0250 36966-1000
TS36966-0050EA 108 USD InStock
TS36966-0050 TS36966-0250 TS36966-1000
Di-tert-butyl iminodicarboxylate 97%
Di-tert-butyl iminodicarboxylate


Formula: C₁₀H₁₉NO₄
Melting Pt: 114…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043309
CAS Number: 51779-32-9
Order Now
Specification Test Results
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty